Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiDry™ ATG™ Ultra Lightweight Nitrile Coated Gloves

Catalog No. 19148459 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-459 X-Large
19-148-456 Small
19-148-457 Medium
19-148-458 Large
19-148-460 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-148-459 Supplier PIP™ Supplier No. 56424/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ultra thin design is comfortable and flexible, while also ensuring maximum fingertip tactile sensitivity. GripTech™ coating offers non-slip grip capabilities in wet or oily applications.

  • Lightweight nitrile coating on a fine gauge seamless knit nylon liner
  • Ultra thin design is comfortable and flexible, while also ensuring maximum finger tip tactile sensitivity
  • Exclusive GripTech™ coating offers non-slip grip capabilities in wet or oily applications
  • LiquiTech™ Barrier Technology provides resistant to oils and liquids
  • Locking cuff ensures the glove doesn't slip off the hand

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) Assembly, Precision Handling, and General Maintenance
Glove Cuff Color Purple
Glove Cuff Style Continuous Knit
Series 56-424
Yarn Gauge 15
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.