Learn More
Medicom™ SafeMask™ SofSkin™ Face Masks
Catalog No. 19910708 Shop All Medicom Products
Click to view available options
Color:
Blue
Lavender
Pink
White
ASTM Level:
ASTM Level 1
ASTM Level 3
This item is not returnable. View return policy
This item is not returnable. View return policy
SofSkin™ procedural face masks with earloops are ultra-soft, lightweight, non-irritating and remain cool on skin.
  • Unique cellulose-blend inner layer is ultra-soft, white, dye-free
  • Choose from ASTM Level 1 or 3
  • Adjustable nose piece
  • Unique shingle design
  • Soft, stretchy, latex-free, sonically sealed earloops
  • Comfortable and non-irritating
  • Customized fit and breathability
  • Keeps skin clean and dry
  • Will not lint, tear or shred
  • Easy removal to help prevent self-contamination
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model 2087
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.