Learn More
PIP™ Eva™ Petite Women's Pink Safety Glasses
Catalog No. 19823994 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Clear, Pink
Lens Coating:
Anti-fog, Scratch-resistant
Hardcoat
Lens Tint:
Clear
Gray
Pink
Temple Color:
Clear/Pink
Gray, Pink
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint Temple Color
19-823-994 Clear, Pink Hardcoat Clear Clear/Pink
19-823-995 Clear, Pink Anti-fog, Scratch-resistant Clear Clear/Pink
19-823-997 Clear, Pink Hardcoat Gray Gray, Pink
19-823-996 Clear, Pink Hardcoat Pink Clear/Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-823-994 Supplier PIP™ Supplier No. 250110900
Add to Cart
Add to Cart
This item is not returnable. View return policy
smaller and more narrow than standard safety glasses
Feature a Pink Ribbon on frames; a portion of the proceeds from the purchase of this product will go to help breast cancer research.
  • Polycarbonate lenses block 99.9% of UV A/B radiation
  • Smaller vertical size fits on a woman's higher cheekbones
  • Soft rubber temples offer a comfortable and secure fit
  • Lightweight construction ensures all-day comfort
  • Rimless frame, rubber temple ends, polycarbonate bridge
  • Base Curve: 11
Compliance
Meets ANSI™ Z87+ high-impact standards.
Specifications
Lens Coating Hardcoat
Certifications/Compliance Meets ANSI Z87+ high-impact standards.
Lens Tint Clear
Product Type Women's Rimless Safety Glasses
For Use With (Application) Used for general manufacturing
Frame Color Clear, Pink
Temple Style Wraparound
Temple Color Clear/Pink
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.