Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Seamless Knit Nylon Gloves with Nitrile Coated MicroSurface Grip

Catalog No. 19130344C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-344C Medium
19-130-344A X-Small
19-130-344B Small
19-130-344D Large
19-130-344F 2X-Large
19-130-344E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-344C Supplier PIP™ Supplier No. 34400/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit shell increases comfort and finger dexterity, while the MicroSurface coating is liquid impermeable to enhance grip in dry or oily conditions.

  • Seamless knit shell increases comfort, finger dexterity, and breathability
  • Nitrile MicroSurface coating is impermeable to provide an elevated grip in dry and oily conditions
  • Coating is infused with thousands of tiny suction cup pockets that create a vacuum effect to dispense fluids away on contact
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Resistant to chemicals, water, and ultraviolet light
  • Washable

Specifications

ANSI Abrasion Rating ANSI 1
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Black, Green
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
Style Standard
For Use With (Application) Oily Parts Assembly, Metal Handling, Fishing, and Machine Operation
Glove Cuff Style Knit
Series 34-400
Yarn Gauge 18
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.