Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Seamless Knit Nylon Gloves with Nitrile MicroSurface Coating

Catalog No. 19153161 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-153-161 X-Small
19-153-162 Small
19-153-163 Medium
19-153-164 Large
19-153-165 X-Large
19-153-166 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-153-161 Supplier PIP™ Supplier No. 34500/XS
This item is not returnable. View return policy

A seamless knit shell offers increased comfort , dexterity, and breathability. The MicroSurface coating is impermeable to provide enhanced grip capabilities in dry and oily conditions.

  • Thousands of tiny suction cup pockets create a vacuum effect that dispenses fluids away when they come in contact with the microsurface coating
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Gloves are washable, resistant to chemicals, water, and ultraviolet light
  • Color coded hems for easy size identification
  • Ideal for handling/assembling parts and materials, equipment maintenance, fastening, and anchoring

Specifications

Construction Seamless Knit
Coating Material Nitrile
Color Black, Blue
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
EN388 Standard 4121X
For Use With (Application) Part Handing/Assembly, Equipment Maintenance, Fastening, and Anchoring
Glove Cuff Style Knit
Hem Color Purple
Series 34-500
Yarn Gauge 13
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.