Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Premium Grade Crinkle Latex Coated Cotton/Polyester Gloves

Catalog No. 19120161 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-161 X-Large
19-151-289 Small
19-816-906 Medium
19-816-905 Large
19-151-290 2X-Large
19-151-288 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-120-161 Supplier PIP™ Supplier No. 39C1300/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Latex crinkle coatings feature a surface texture that is designed to channel fluids away from the glove, allowing for better contact on wet and dry surfaces.

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern
  • Color coded hems for easy size identification
  • Ideal for handling/cleanup of abrasive parts and materials, demolition, masonry, metal fastening, and assembly

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Natural, Blue
Exterior Finish Fully Textured
Grade Premium
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) Handling/Cleanup of Abrasive Materials, Demolition, Masonry, Tile Work, and Metal Fastening
Glove Cuff Style Knit
Series 39-C1300
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.