Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP Smooth Nitrile Coated Seamless Knit Nylon Gloves

Catalog No. 191303719C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-3719C Large
19-130-3719A Small
19-130-3719B Medium
19-130-3719D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-3719C Supplier PIP™ Supplier No. 34225/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit nylon shell increases comfort, finger dexterity, and breathability. Smooth nitrile coating is highly impermeable to liquids for enhanced grip capabilites in applications that require secure holding power.

  • Solid dipped nitrile coating enhances grip capabilities, and is impermeable to liquids
  • Knit wrist helps to prevent dirt and debris from entering the glove
  • Breathable nylon back increases breathability and comfort
  • Ideal for wet or dry applications that require secure holding power

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Large
Style Standard
For Use With (Application) Manufacturing/Assembly Applications Where Abrasion, Cut and Puncture Resistance is Required
Glove Cuff Style Continuous Knit
Series 34-225
Yarn Gauge 13
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.