Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ G-Tek™ GP Smooth Nitrile Coated Seamless Knit Nylon Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Solid dipped nitrile coating enhances grip capabilities, and is impermeable to liquids
- Knit wrist helps to prevent dirt and debris from entering the glove
- Breathable nylon back increases breathability and comfort
- Ideal for wet or dry applications that require secure holding power
Specifications
Specifications
Coated Area | Palm, Fingers |
Construction | Seamless Knit |
Coating Material | Nitrile |
Color | White |
Exterior Finish | No Texture |
Hand | Specific |
Material | Nylon |
Product Type | Seamless Knit Gloves |
Size | Medium |
For Use With (Application) | Manufacturing/Assembly Applications where Abrasion, Cut and Puncture Resistance is Required |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction