Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ NPB2 Heavy Weight Nylon Gloves with Extra Thick Polyurethane Coating

Catalog No. 19175104A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-175-104A Small
19-175-104B Medium
19-175-104C Large
19-175-104D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-175-104A Supplier PIP™ Supplier No. 33325/S
This item is not returnable. View return policy

A heavy weight, seamless knit nylon shell increases comfort, finger dexterity, and breathability. The extra thick polyurethane coating provides puncture and abrasion resistance, while also enhancing tactile sensitivity.

  • Heavy weight, seamless knit nylon shell increases comfort, finger dexterity, and breathability
  • Extra thick polyurethane coating takes on the surface properties of the glove liner
  • Provides a tactile grip in dry and slightly oily conditions
  • Ideal for applications that require dexterity for precise handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Breathable nylon back increases user comfort
  • Packed in pairs for ease of distribution

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Black
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
Style Standard
For Use With (Application) Electronic/Computer Assembly, Quality Control, and Inspection
Glove Cuff Style Continuous Knit
Series 33-325
Length (English) 8.5 in.
Length (Metric) 21.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.