Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ BN Smooth Nitrile Coated Seamless Knit Nylon Gloves

Catalog No. 191301181B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1181B Medium
19-130-1181C Large
19-130-1181D X-Large
19-130-1181A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1181B Supplier PIP™ Supplier No. 34C229/M
This item is not returnable. View return policy

Smooth nitrile coating is highly impermeable to liquids to provide a tacky dry grip for applications that require secure holding power.

  • Seamless knit nylon shell increases comfort, finger dexterity, and breathability
  • Nitrile coating is highly impermeable to liquids
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Launderable

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color White, Blue
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
EN388 Standard 4131
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 34-C229
Yarn Gauge 13
Length (English) 9.4 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.