Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Black Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. 19149608 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-608 X-Small
19-149-609 Small
19-149-613 2X-Large
19-149-610 Medium
19-149-612 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-149-608 Supplier PIP™ Supplier No. 33B125/XS
This item is not returnable. View return policy

Seamless knit nylon construction offers increased comfort, dexterity, and breathability. The polyurethane grip provides a tactile grip in dry, and slightly oily conditions.

  • Seamless knit nylon shell offers comfort, finger dexterity, and breathability
  • Polyurethane coating provides a tactile grip in dry and slightly oily conditions
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ideal for applications that require dexterity, and precise handling
  • Breathable back for comfort
  • Packed in pairs for ease of distribution

Specifications

Construction Seamless Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A1
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
EN388 Standard 4131
For Use With (Application) Electronic Assembly, Quality Control, Inspection, and General Assembly
Glove Cuff Style Continuous Knit
Hem Color Purple
Series 33-B125
Yarn Gauge 13
Length (English) 7.5 in.
Length (Metric) 19 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.