Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Touch Seamless Knit Polyurethane Coated Nylon/Polyester Gloves

Catalog No. 19149900 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-900 X-Small
19-149-901 Small
19-149-903 Large
19-149-904 X-Large
19-149-905 2X-Large
19-149-902 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-149-900 Supplier PIP™ Supplier No. 33GT125/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Static wired thumb, index finger, and middle finger allows users to operate touchscreen phones or devices without removing their gloves.

  • Static wired thumb, index finger and middle finger allows users to operate touchscreen phones or devices without removing gloves
  • Polyurethane coatings take on the surface properties of the glove liner to provide a tactile grip in dry and slightly oily conditions
  • Seamless knit construction is ideal for applications that require dexterity for precise handling
  • Breathable back enhances user comfort

Specifications

ANSI Abrasion Rating ANSI 1
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) Construction, Sanitation, Quality Control, Inspection, and General Assembly
Glove Cuff Style Continuous Knit
Series 33-GT125
Yarn Gauge 13
Length (English) 8.3 in.
Length (Metric) 21.1 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.