Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Premium Grade Top Grain Goatskin Leather Drivers Gloves

Catalog No. 191201538A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-1538A Small
19-120-1538B Medium
19-120-1538C Large
19-120-1538D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-1538A Supplier PIP™ Supplier No. 713618/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Top grain goatskin leather construction is soft, abrasion-resistant, and highly recommended for applications that require tactile sensitivity.

  • Top grain leather is smooth, provides durability and dexterity
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification
  • Meets the content requirements for Chromium VI free leather

Specifications

Color Natural
Grade Premium
Hand Specific
Material Top Grain Goatskin Leather
Product Type Driver Gloves
Size Small
Style Standard
For Use With (Application) Construction, Machine Operation, Utility Work and General Applications
Glove Cuff Style Open
Hem Color Red
Series 71-3618
Thumb Style Keystone
Length (English) 8.8 in.
Length (Metric) 22.3 cm
Weight 19 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.