Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Medium Weight Seamless Knit Cotton / Polyester Half-Finger Gloves

Used for warehousing, parts handling, assembly and food processing.

Manufacturer:  PIP™ 35C119/L

 View more versions of this product

Catalog No. 19-121-231



Add to cart

Description

Description

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows replacing one glove at a time, rather than pairs
  • Half-finger for greater dexterity, sensitivity and breathability
  • Can be used as a glove liner
  • Small size fits ladies well
  • Made from component materials which comply with Federal regulations for food contact

Specifications

Specifications

7.7 x 3.9 in. (19.5 x 9.8cm)
Continuous knit
Large
9.8cm
7.7 in.
Fingerless
Cotton, Polyester
3.9 in.
Natural
19.5cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit