Learn More
PIP™ Kut-Gard™ Chemical-Resistant Gloves
Catalog No. 19100927 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-100-927 Medium
19-100-926 Small
19-100-928 Large
19-100-929 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-100-927 Supplier PIP™ Supplier No. 12350M
This item is not returnable. View return policy
Specifications
Certifications/Compliance OSHA CFR 1910.269
Material Nuaramid
Size Medium
Thickness (English) 7 mil
Thickness (Metric) 0.17 mm