Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Kut-Gard™ Dyneema™ Antimicrobial Cut-Resistant Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Blue
Green
White
Yellow
Length (English):
8.3 in.
10 in.
11 in.
11.5 in.
12 in.
Length (Metric):
21 cm
25.4 cm
27.9 cm
29.2 cm
30.5 cm
Description
- Seamless knit construction provides comfort, fit, and dexterity
- Dyneema™ blend enhances the gloves performance without sacrificing flexibility or dexterity and ensures maximum protection
- Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
- Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Good gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Antimicrobial gloves are non-toxic and non-corrosive and complies with FDA, EPA, NSF and USDA standards for food contact
- Dyneema is 15 times stronger than steel on a weight for weight basis - a High Performance Polyethylene (HPPE) fiber made from Ultra-High Molecular Weight Polyethylene (UHMWPE)
- Dyneema fiber is much thinner and more flexible than traditional materials, such as leather and cotton, while offering higher protection
Specifications
Specifications
Certifications/Compliance | FDA 21CFR 170-199 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | Gray |
Cut Resistance Rating | ANSI A5 |
Hand | Ambidextrous |
Material | Dyneema, Stainless Steel Fiber, Synthetic Fiber |
Product Type | Cut-resistant Gloves |
Size | Large |
For Use With (Application) | Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction