Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Dyneema™ Antimicrobial Cut-Resistant Gloves

Catalog No. 19149604 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Blue
Green
White
Yellow
Length (English):
8.3 in.
10 in.
11 in.
11.5 in.
12 in.
Length (Metric):
21 cm
25.4 cm
27.9 cm
29.2 cm
30.5 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color Length (English) Length (Metric)
19-149-604 Small Yellow 10 in. 25.4 cm
19-149-603 X-Small Black 8.3 in. 21 cm
19-149-605 Medium Blue 11 in. 27.9 cm
19-149-606 Large White 11.5 in. 29.2 cm
19-149-607 X-Large Green 12 in. 30.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-149-604 Supplier PIP™ Supplier No. 22900S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium-weight ANSI Level 7 cut-resistant gloves are ideal for any application where sharp cutting instruments are used.

  • Seamless knit construction provides comfort, fit, and dexterity
  • Dyneema™ blend enhances the gloves performance without sacrificing flexibility or dexterity and ensures maximum protection
  • Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
  • Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
  • Can be worn on its own or as a glove liner
  • Good gripping properties
  • Shrink resistant
  • Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
  • Antimicrobial gloves are non-toxic and non-corrosive and complies with FDA, EPA, NSF and USDA standards for food contact
  • Dyneema is 15 times stronger than steel on a weight for weight basis - a High Performance Polyethylene (HPPE) fiber made from Ultra-High Molecular Weight Polyethylene (UHMWPE)
  • Dyneema fiber is much thinner and more flexible than traditional materials, such as leather and cotton, while offering higher protection

Specifications

Certifications/Compliance FDA 21CFR 170-199
Construction Seamless Knit
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Dyneema, Stainless Steel Fiber, Synthetic Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Hem Color Yellow
Model 22-900S
Series Kut-Gard™ 22-900
Yarn Gauge 7
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.