Learn More
PIP™ Kut-Gard™ Dyneema™ Antimicrobial Cut-Resistant Gloves
Shop All Protective Industrial Products Inc. ProductsDescription
- Seamless knit construction provides comfort, fit, and dexterity
- Dyneema™ blend enhances the gloves performance without sacrificing flexibility or dexterity and ensures maximum protection
- Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
- Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Good gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Antimicrobial gloves are non-toxic and non-corrosive and complies with FDA, EPA, NSF and USDA standards for food contact
- Dyneema is 15 times stronger than steel on a weight for weight basis - a High Performance Polyethylene (HPPE) fiber made from Ultra-High Molecular Weight Polyethylene (UHMWPE)
- Dyneema fiber is much thinner and more flexible than traditional materials, such as leather and cotton, while offering higher protection
Specifications
Specifications
| Certifications/Compliance | FDA 21CFR 170-199 |
| Construction | Seamless Knit |
| Coating Material | Uncoated |
| Color | Gray |
| Cut Resistance Rating | ANSI A5 |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Dyneema, Stainless Steel Fiber, Synthetic Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.