Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Dyneema™ Cut-Resistant Gloves

Catalog No. 19100931 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-100-931 X-Small Blue
19-100-930 2X-Small -
19-100-932 Small White
19-100-933 Medium Red
19-100-934 Large Yellow
19-100-935 X-Large Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-100-931 Supplier PIP™ Supplier No. 17D200/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight uncoated gloves provide dexterity, tactile sensitivity and cut resistance. Ideal for use as a glove liner.

  • 100% Dyneema
  • Lightweight
  • Seamless construction
  • Can be used as a glove liner
  • Color coded cuff hems for easy size identification
  • Dyneema is a brand of polyethylene fibers that are 10 to 15 times stronger than steel
  • More durable than polyester and a strength that is 40% greater than aramid fiber

Specifications

Certifications/Compliance ASTM F2992-15 ANSI Cut Level A2; ASTM F1790-05 ANSI Cut Level 2; 21CFR 170-199
Construction Seamless Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Dyneema
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Food processing
Glove Cuff Color Blue
Glove Cuff Style Continuous Knit
Model 17-D200/XS
Series Kut-Gard™ 17-D200
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.