Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Dyneema™ Cut-Resistant Gloves

Catalog No. p-4379673
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Blue
Green
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight uncoated gloves provide dexterity, tactile sensitivity and cut resistance. Ideal for use as a glove liner.

  • 100% Dyneema
  • Lightweight
  • Seamless construction
  • Can be used as a glove liner
  • Color coded cuff hems for easy size identification
  • Dyneema is a brand of polyethylene fibers that are 10 to 15 times stronger than steel
  • More durable than polyester and a strength that is 40% greater than aramid fiber

Specifications

Certifications/Compliance ASTM F2992-15 ANSI Cut Level A2; ASTM F1790-05 ANSI Cut Level 2; 21CFR 170-199
Construction Seamless Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Material Dyneema
Product Type Cut-resistant Gloves
Size 2X-Small
For Use With (Application) Food processing
Glove Cuff Style Continuous Knit
Model 17-D200/XXS
Series Kut-Gard™ 17-D200
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.