Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ PolyKor™ Seamless Knit Antimicrobial Gloves

Catalog No. 191304478C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Length (English):
8 in.
8.5 in.
10 in.
10.5 in.
11 in.
Length (Metric):
20.3 cm
21.6 cm
25.4 cm
26.7 cm
27.9 cm
Size:
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Blue
Green
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Size Glove Cuff Color
19-130-4478C 10 in. 25.4 cm Medium Blue
19-130-4478A 8 in. 20.3 cm X-Small Black
19-130-4478B 8.5 in. 21.6 cm Small Yellow
19-130-4478D 10.5 in. 26.7 cm Large White
19-130-4478E 11 in. 27.9 cm X-Large Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-4478C Supplier PIP™ Supplier No. 22720M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium-weight antimicrobial cut-resistant gloves are ideal for any application where sharp cutting instruments are used.

  • Seamless knit construction provides comfort, fit, and dexterity
  • PolyKor™ engineered yarns offer exceptional cut and slash resistance
  • Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
  • Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
  • Can be worn on its own or as a glove liner
  • Good gripping properties
  • Shrink resistant
  • Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
  • Products constructed with PolyKor™ engineered yarn are suitable for protection against mechanical hazards in the toughest environments
  • PolyKor products are highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection

Specifications

Certifications/Compliance FDA 21 CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Stainless Steel Fiber, Synthetic Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling
Glove Cuff Color Blue
Glove Cuff Style Elasticated
Hem Color Blue
Model 22-720M
Series Kut Gard™ 22-720
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.