Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Superior Grade Top Grain Cowhide Leather Drivers Gloves with Hi-Vis Fingertips

Catalog No. 19013373A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013-373A X-Small
19-013-373B Small
19-013-373C Medium
19-013-373D Large
19-013-373E X-Large
19-013-373F 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-013-373A Supplier PIP™ Supplier No. 68165HV/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature highly visible coloring on the finger tips, with WATCH YOUR HANDS printed on the back of the palm to keep workers alert at all times.

  • Top grain cowhide leather construction provides comfort, durability, abrasion resistance, and breathability
  • Top Grain leather is smooth, provides durability and dexterity
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification
  • Meets the content requirements for Chromium VI Free Leather

Specifications

Color Natural, Hi-vis Orange
Grade Superior
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size X-Small
Style Standard
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Light Blue
Series 68-165HV
Thumb Style Keystone
Length (English) 8.7 in.
Length (Metric) 22 cm
Weight 27.7 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.