Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Elite™ Seamless Knit Nylon Gloves with Nitrile Coating

Catalog No. 19153130 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-153-130 X-Small
19-153-132 Medium
19-153-133 Large
19-153-134 X-Large
19-153-131 Small
19-153-135 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-153-130 Supplier PIP™ Supplier No. 34274/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

MaxiFlex™ Elite™ gloves are 35% thinner and 40% more breathable than traditional foam nitrile gloves. MicroFoam coating is compatible with light oils for enhanced grip capabilities in all types of conditions.

  • Knit wrist helps prevent dirt and debris from entering the glove
  • 30% thinner than traditional foam nitrile gloves while still offering similar mechanical performance to traditional foam nitrile gloves
  • Heat created by the hand is quickly eliminated through the breathable fabric
  • Patented micro-foam nitrile coating offers 360° breathability
  • Micro-cup finish allows for a controlled grip in dry, or slightly oily conditions
  • Ultra light liner produces smooth and rounded fingertips to maximize sensitivity
  • Glove fit mimics the hand at rest, reducing hand fatigue and increasing comfort
  • Penetration-free liner has a soft feel when resting against the skin
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • 100% Silicone free

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Nitrile
Color Blue
Exterior Finish Fully Textured
Hand Specific
Material Nylon, Lycra
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
For Use With (Application) Part Handing/Assembly, Equipment Maintenance, Fastening, and Anchoring
Glove Cuff Color Dark Blue
Glove Cuff Style Continuous Knit
Hem Color Blue
Series 34-274
Yarn Gauge 18
Length (English) 7.7 in.
Length (Metric) 19.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.