Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Elite™ Seamless Knit Nylon Gloves with Nitrile Coating & Micro-Dotted Palms

Catalog No. 19153141 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-153-141 X-Small
19-153-142 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-153-141 Supplier PIP™ Supplier No. 34244/XS
This item is not returnable. View return policy

Gloves feature a micro-foam nitrile coating for enhanced grip capabilities in dry, or light oil applications. Raised micro-dots on the palm increase durability and cushioning during highly repetitive tasks.

  • Micro-Foam nitrile coating is compatible with light oils to provide enhanced grip and abrasion resistance
  • Raised micro-dots increase durability and offer additional cushioning in highly repetitive applications that require precision handling
  • Gloves are 35% thinner than traditional foam gloves for 40% better breathability
  • Patented micro-foam nitrile coating offers 360° breathability
  • Ultra light liner produces smooth, rounded fingertips to maximize sensitivity
  • Design mimics the hand at rest to reduce hand fatigue
  • Penetration free liner has a soft feel when resting against the skin
  • Gloves are laundered prior to final packaging to ensure they are clean and safe for use

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Nitrile
Color Blue
Hand Specific
Material Nylon, Lycra
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) General Assembly, Automotive Components, Micro-Engineering, and Warehousing
Glove Cuff Style Continuous Knit
Hem Color Blue
Palm Type Raised Micro-Dots
Series 34-244
Yarn Gauge 18
Length (English) 7.7 in.
Length (Metric) 19.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.