Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFoam™ Lite Seamless Knit Nylon Gloves with Foam Nitrile Coating

Catalog No. 19160302 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-302 X-Small
19-160-303 Small
19-160-304 Medium
19-160-306 Large
19-160-307 X-Large
19-160-308 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-160-302 Supplier PIP™ Supplier No. 34900/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam nitrile coating is compatible with light oils to enhance grip and provide abrasion resistance. Seamless nylon shell increases comfort, finger dexterity, and breathability.

  • Foam nitrile coating is compatible with light oils to enhance grip and provide abrasion resistance without sacrificing dexterity
  • Seamless nylon shell increases comfort, finger dexterity, and breathability
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Surface of the glove designed to channel oil away to give a better grip
  • Gloves laundered prior to final packaging ensuring that they are clean and safe for use
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Designed to produce minimal lint and dust
  • Dark color combination hides dirt for prolonged glove life
  • 100% silicone free

Specifications

ANSI Abrasion Rating ANSI 1
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Painting, and Horticulture
Glove Cuff Color Gray
Glove Cuff Style Continuous Knit
Series 34-900
Yarn Gauge 15
Length (English) 7 in.
Length (Metric) 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.