Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Leather Palm Black Mechanics Gloves

Catalog No. 19823942 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-942 Large
19-100-526 Small
19-823-941 Medium
19-823-943 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-823-942 Supplier PIP™ Supplier No. 120MX2805/L
This item is not returnable. View return policy

Designed to protect the professional mechanic or enthusiast from heat, cold, cuts, and abrasions.

  • Synthetic leather palms and thumbs provide dexterity and grip in dry, or slightly oily applications
  • Spandex back provides a snug fit for increased grip & finger control
  • Double stitched to provide additional durability
  • Hook & Loop closure for easy on/off and a secure fit
  • Launderable for extended life and to reduce replacement costs
  • Ideal for automotive, aerospace, or general industrial maintenance

Specifications

Back Material Spandex
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Large
For Use With (Application) Automotive Assembly, Aerospace, or General Industrial Maintenance
Glove Cuff Style Hook and Loop
Series 120-MX2805
Length (English) 9 in.
Length (Metric) 22.8 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.