Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ NOVAX™ Class 4 Black Natural Rubber Electrical Insulating Gloves
Click to view available options
Size:
9
10
11
12
Length (English):
16 in.
18 in.
Description
- Class 4 electrical insulating gloves constructed of natural rubber
- Flexible material allows workers to easily handle small, delicate parts
- Contoured shape is comfortable and helps to reduce hand fatigue
- Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
- Ideal for use around electrical and arc flash hazards
- Individually numbered for traceability
- Stamped with factory manufacturing date
- Silicone-free
Specifications
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 4, Type I, NFPA 70E |
Applications | Industrial |
Color | Black, Orange |
Exterior Finish | No Texture |
Material | Natural Rubber |
Size | 10 |
For Use With (Application) | Electrical Utility Work, Hybrid Automobiles, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Series | 155-4-16 |
Length (English) | 16 in. |
Show More |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction