Learn More
PIP™ NOVAX™ Class 4 Black Natural Rubber Electrical Insulating Gloves
Shop All Protective Industrial Products Inc. ProductsDescription
- Class 4 electrical insulating gloves constructed of natural rubber
- Flexible material allows workers to easily handle small, delicate parts
- Contoured shape is comfortable and helps to reduce hand fatigue
- Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
- Ideal for use around electrical and arc flash hazards
- Individually numbered for traceability
- Stamped with factory manufacturing date
- Silicone-free
Specifications
Specifications
| Certifications/Compliance | ANSI/ASTM D120, Class 4, Type I, NFPA 70E |
| Applications | Industrial |
| Color | Black, Orange |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Natural Rubber |
| Product Type | Electrical Insulating Gloves |
| Size | 11 |
| For Use With (Application) | Electrical Utility Work, Hybrid Automobiles, Industrial Plant Maintenance, and Power Generation |
| Glove Cuff Style | Straight |
| Show More |
WARNING: Cancer - www.P65Warnings.ca.gov
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.