Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Polyester Gloves with Double-Sided PVC Grip

Catalog No. 19121186B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-186B Small
19-121-186D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-121-186B Supplier PIP™ Supplier No. 393013/S
This item is not returnable. View return policy

Gloves feature a PVC coating on both sides in a honeycomb criss-cross pattern for enhanced grip capabilities.

  • Seamless knit construction is comfortable and flexible
  • Polyester shell dries fast in wet applications
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Double-sided PVC coating in a honeycomb criss-cross pattern enhances grip, as well as cut and abrasion resistance

Specifications

Construction Seamless Knit
Coating Material PVC
Color Orange
Material Polyester
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Fishing, Glass/Plywood/Lumber Handling
Glove Cuff Style Continuous Knit
Series 39-3013
Yarn Gauge 10
Length (English) 9.7 in.
Length (Metric) 24.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.