Learn More
PIP™ PowerGrab™ Plus Latex Coated Cotton/Polyester Work Gloves
Catalog No. 19149629 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-629 Large
19-149-626 X-Small
19-149-628 Medium
19-149-630 X-Large
19-149-627 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-149-629 Supplier PIP™ Supplier No. 39C1600/L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex microsurface coating enhances grip capabilities in wet or dry conditions by acting like tiny suction cups that attach themselves firmly to the material being handled.
  • Latex microsurface coating provides a secure grip in dry and wet conditions
  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern
  • Breathable back for comfort
Specifications
Construction Seamless Knit
Coating Material Latex
Color Blue, Gray
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Polycotton
Product Type Dip-coated Gloves
Size Large
EN388 Standard 3242
Glove Cuff Style Knit
Length (English) 10 in.
Length (Metric) 25.5 cm