Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Premium Grade Top Grain Pigskin Leather Drivers Gloves

Catalog Number 19816843
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Top grain pigskin leather construction provides breathability due its porous texture, becomes softer with use, and withstands moisture without stiffening.

  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Pigskin will return to its original pliability even after being soaked in water
  • Elastic closure sewn inside glove for safety and comfort
  • Consistent grading and superior workmanship
  • Color coded hems for easy size identification
  • Ideal for construction, machine operation, utility work, and many other general applications
Color Natural
Grade Premium
Hand Specific
Material Top Grain Pigskin Leather
Product Type Driver Gloves
Size Small
For Use With (Application) Construction, Machine Operation, Utility Work, and General Applications
Glove Cuff Style Open
Hem Color Red
Series 70-368
Thumb Style Keystone
Length (English) 8.8 in.
Length (Metric) 22.3 cm
Weight 24 lb. (Case)
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit