Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Polyurethane-Coated Nylon Clean Environment Gloves

Catalog No. 19816902 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-902 Large
19-816-903 Medium
19-120-1485D X-Large
19-816-904 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-816-902 Supplier PIP™ Supplier No. 40C125/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit nylon shell is comfortable and reduces hand fatigue, while the polyurethane coating provides a secure grip in dry, or slightly wet conditions.

  • Knit wrist helps prevent dirt and debris from entering the glove
  • Constructed from nylon filament yarns for low lint applications
  • Nylon yarns absorb perspiration to leave hands cool and comfortable
  • Can be worn as a liner under disposable and other liquid-proof gloves
  • Color coded hems for easy size identification

Specifications

Coated Area Fingertips
Construction Seamless Knit
Coating Material Polyurethane
Color White
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Product Handling, Electronics Assembly, Photography Labs, and Computer Manufacturing
Glove Cuff Color Green
Glove Cuff Style Knit
Series 40-C125
Length (English) 9.4 in.
Length (Metric) 24 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.