Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ XtraTuff™ Oil Resistant PVC Gloves with 25 in. Extended PVC Sleeves

Catalog No. 19175110 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-175-110 Medium
19-175-111 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-175-110 Supplier PIP™ Supplier No. 588657/M
This item is not returnable. View return policy

Heat-sealed PVC sleeves are reinforced to prevent tearing and punctures, with an elastic collar for forearm protection in non-chemical environments. Ideal for applications that require a combination of liquid and cut protection.

  • Rough coated palm enhances wet and dry grip capabilities
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Oil and liquid-resistant, staying soft and strong even after exposure to machine oils
  • Heat-sealed PVC sleeves are reinforced to prevent tearing and punctures
  • Elastic collar provides forearm protection in non-chemical environments
  • Ideal for applications where machine oil is present
  • Liquid and cut protection combined into one glove

Specifications

Color Blue, Clear
Exterior Finish Fully Textured
Hand Specific
Material PVC
Product Type Dip-coated Gloves
Size Medium
EN388 Standard 4121
EN ISO 374-1 CHEMICAL LETTER CODES AJK
For Use With (Application) Machine Oils, Liquid/Cut Protection
Glove Cuff Style Elasticated
Liner Material Cotton
Series 58-8657
Length (English) 25 in.
Length (Metric) 63.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.