Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP4K2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515285
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515285 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515285 Supplier Invitrogen™ Supplier No. PA515285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology.

PIP4K2A lipid kinase or phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, is a member of the phosphatidylinositol-5-phosphate 4-kinase family that catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-4,5-bisphosphate. PIP4K2A plays as essential role in the phosphoinositide signal transduction cascades as the precursor to second messengers and is involved in the regulation of secretion, cell proliferation, differentiation, and motility. PIP4K2A may be one of the factors related to the regulation of the beta-globin gene expression and the different levels of Hb H in alpha-thalassemic patients. Diseases associated with PIP4K2A include B-Lymphoblastic Leukemia/Lymphoma With Hyperdiploidy and Bipolar Disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP4K2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PIP4K2A
Gene Accession No. O70172, P48426
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-4-phosphate-5-kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-5-phosphate 4-kinase A; AW742916; diphosphoinositide kinase 2-alpha; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-4-phosphate 5-kinase, type II, alpha; phosphatidylinositol-5-phosphate 4-kinase type 2 alpha; phosphatidylinositol-5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-5-phosphate 4-kinase, type II, alpha; PI(5)P 4-kinase type II alpha; PI5P4KA; Pip4k2a; PIP4KII-alpha; PIP5K2; Pip5k2a; PIP5KIIA; PIP5KIIalpha; PIP5KII-alpha; PIP5KIII; PIPK; PIPK2 alpha; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; ptdIns(5)P-4-kinase isoform 2-alpha; type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Gene Symbols PIP4K2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 107-139 amino acids from the N-terminal region of human PIP4KII-alpha (PIP4K2A).
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18718, 5305
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.