Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP4K2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566471 Supplier Invitrogen™ Supplier No. PA566471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDSPGNLLSFP RFFGPGEFDP SVDVYAMKSH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP4K2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIP4K2B
Gene Accession No. P78356
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; AI848124; c11; diphosphoinositide kinase 2-beta; LOW QUALITY PROTEIN: phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type 2 beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP4KII-beta; Pip5k2b; PIP5KIIB; PIP5KIIbeta; PIP5P4KB; PIPKIIgamma; PTDINS(4)P-5-kinase; ptdIns(5)P-4-kinase isoform 2-beta
Gene Symbols PIP4K2B
Host Species Rabbit
Immunogen Recombinant Human PIP4K2B. Recombinant protein control fragment (Product #RP-105227).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8396
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.