Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP4K2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576328 Supplier Thermo Scientific Supplier No. PA576328
Add to Cart
Add to Cart

PIP4K2B Polyclonal Antibody

PIP4K2B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP4K2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PIP4K2B
Gene Accession No. O88377, P78356, Q80XI4
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; AI848124; c11; diphosphoinositide kinase 2-beta; LOW QUALITY PROTEIN: phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type 2 beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP4KII-beta; Pip5k2b; PIP5KIIB; PIP5KIIbeta; PIP5P4KB; PIPKIIgamma; PTDINS(4)P-5-kinase; ptdIns(5)P-4-kinase isoform 2-beta
Gene Symbols PIP4K2B
Host Species Rabbit
Immunogen Recombinant protein of human PIP4K2B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108083, 8396, 89812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.