Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP4K2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596902 Supplier Invitrogen™ Supplier No. PA596902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSNCTSTTA VAVAPLSASK TKTKKKHFVC QKVKLFRASE PILSVLMWGV NHTINELSNV PVPVMLMPDD FKAYSKIKVD NHLFNKENLP SRFKFKEYCP MVFRNLRERF GIDDQDYQNS VTRSAPINSD SQGRCGTRFL TTYDRRFVIK TVSSEDVAEM HNILKKYHQF IVECHGNTLL PQFLGMYRLT VDGVETYMVV TRNVFSHRLT VHRKYDLKGS TVAREASDKE KAKDLPTFKD NDFLNEGQKL HVGEESKKNF LEKLKRDVEF; Positive Samples: BT-474, HepG2, A-549; Cellular Location: Cell membrane, Cytoplasm, Endoplasmic reticulum membrane, Nucleus, Peripheral membrane protein.

The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP4K2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIP4K2B
Gene Accession No. P78356
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; AI848124; c11; diphosphoinositide kinase 2-beta; LOW QUALITY PROTEIN: phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type 2 beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP4KII-beta; Pip5k2b; PIP5KIIB; PIP5KIIbeta; PIP5P4KB; PIPKIIgamma; PTDINS(4)P-5-kinase; ptdIns(5)P-4-kinase isoform 2-beta
Gene Symbols PIP4K2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PIP4K2B (NP_0035501).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8396
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.