Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP4K2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119828 Supplier Invitrogen™ Supplier No. PA5119828
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: MCF-7 cell lysate, A549 cell lysate, rat cerebellum tissue lysate, mouse brain tissue lysate. Subcellular Location: Cytoplasm, Endoplasmic reticulum.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP4K2C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PIP4K2C
Gene Accession No. O88370, Q8TBX8, Q91XU3
Gene Alias phosphatidylinositol 4-phosphate 5-kinase; phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type 2 gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-5-phosphate 4-kinase, type II, gamma; PI(5)P 4-kinase type II gamma; Pip4k2c; PIP4KII-gamma; PIP5K2C
Gene Symbols PIP4K2C
Host Species Rabbit
Immunogen Recombinant protein within human PIP5K2 gamma aa 100-300.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117150, 140607, 79837
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.