Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP5K1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583084 Supplier Invitrogen™ Supplier No. PA583084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRRASQKERE KPLPTFKDLD FLQDIPDGLF LDADMYNALC KTLQRDCLVL QSFKIMDYSL LMSIHNIDHA QREPLSSETQ YSVDTRRPAP QKALYSTAME SIQGEARRGG TMETDDHMGG IPAR Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.

PIP5K1A lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 alpha phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1 is required for Wnt3a-induced LRP6 phosphorylation at ser1490 in mammalian cells. Wnt3a stimulates the formation of phosphatidylinositol 4,5-bisphosphates through 'frizzled' and 'disheveled', the latter of which directly interacts with and activated PIP5KI. PIP5K1A-mediated PIP(2) production is crucial for HIV-1 entry and the early steps of infection in permissive lymphocytes. Overexpression of wild-type PI4P5K1A increases HIV-1 Env-mediated PIP(2) production and enhances viral replication in primary lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pip5k1a
Gene Accession No. Q99755
Gene Alias 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha; phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1, alpha; phosphatidylinositol-4-phosphate 5-kinase, type I, alpha; PI4P5K-I[a]; PI4P5KIbeta; PI51A; Pip5k1a; PIP5K1-alpha; Pip5k1b; PIP5KIalpha; Pipk5a; ptdIns(4)P-5-kinase 1 alpha
Gene Symbols Pip5k1a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIP5K1A. Recombinant protein control fragment (Product #RP-89191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8394
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.