Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP5K1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545874 Supplier Invitrogen™ Supplier No. PA545874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VFKKIQALKA SPSKKRCNSI AALKATSQEI VSSISQEWKD EKRDLLTEGQ Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

PIP5K1B lipid kinase or phosphatidylinositol-4-phosphate 5-kinase, type I, beta is a member of the phosphatidylinositol-4-phosphate 5-kinase family. PIP5K1B gene contains 17 exons and spans more than 300 kb. The seventeenth exon was found by RT-PCR, and is derived from the 3-prime untranslated region of the PRKACG gene which is located on 9q13 approximately 3 kb downstream of the STM7.I 3-prime untranslated region. The overexpression of PIP5K1B in COS-7 cells induces an increase in short actin fibers and a decrease in actin stress fibers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIP5K1B
Gene Accession No. O14986
Gene Alias MSS4; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type 1 beta; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type I beta; phosphatidylinositol-4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1, beta; phosphatidylinositol-4-phosphate 5-kinase, type I, beta; PI4P5K-I[b]; Pip5k1a; Pip5k1b; PIP5K1-beta; PIP5KIalpha; PIP5KIB2 protein; PIP5KIbeta; Pipk5b; Protein STM-7; ptdIns(4)P-5-kinase 1 beta; ptdIns(4)P-5-kinase beta; STM7; STM-7 protein; Type I phosphatidylinositol 4-phosphate 5-kinase beta; type I phosphatidylinositol-4-phosphate 5-kinase beta
Gene Symbols PIP5K1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PIP5K1B (aa 395-444).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8395
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.