Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K1C Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735725 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5725 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5725 Supplier Proteintech Group Inc Supplier No. 276401AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphatidylinositol-4-phosphate-5-kinase (PIPK) synthesizes phosphatidylinositol-4,5-bisphosphate, which regulates various processes including cell proliferation, survival, membrane trafficking and cytoskeletal organization. The PIPK family is divided into type I, type II and type III. Each type of the PIPK family phosphorylates distinct substrates. They contain an activation loop, which determines their enzymatic specificity and subcellular targeting. The phosphatidylinositol-4-phosphate-5-kinase type I consists of three members, PIPK I alpha, beta and gamma, which are characterized by phosphorylating PI4P on the 5-hydroxyl. PIPK I alpha, designated PIPK I beta in mouse, is expressed in brain tissue. PIPK I beta, designated PIPK I alpha in mouse, is also called STM7. PIPK I gamma has two variants produced by alternative splicing which are expressed in lung, brain and kidneys.

Specifications

Antigen PIP5K1C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PIP5K1C
Gene Accession No. O60331
Gene Alias KIAA0589, LCCS3, PIP5K GAMMA, PIP5K1 gamma, PIP5K1C, PIP5Kgamma, PIP5KIgamma, PtdIns(4)P 5 kinase 1 gamma
Gene Symbols PIP5K1C
Host Species Rabbit
Immunogen PIP5K1C Fusion Protein Ag26525
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23396
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.