Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP5K2 alpha Antibody (OTI3D3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273417MFV500
Description
PIP5K2 alpha Monoclonal antibody specifically detects PIP5K2 alpha in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| PIP5K2 alpha | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 5305 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3D3 | |
| Flow Cytometry | |
| 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-4-phosphate-5-kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha, Diphosphoinositide kinase 2-alpha, EC 2.7.1, EC 2.7.1.149, FLJ13267, phosphatidylinositol-4-phosphate 5-kinase, type II, alpha, Phosphatidylinositol-5-phosphate 4-kinase type II alpha, phosphatidylinositol-5-phosphate 4-kinase type-2 alpha, phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, PI(5)P 4-kinase type II alpha, PIP4KII-alpha, PIP5K2, PIP5K2API5P4KA, PIP5KIIA, PIP5KIIalpha, PIP5KII-alpha, PIP5KIII, PIPK, PtdIns(4)P-5-kinase B isoform, PtdIns(4)P-5-kinase C isoform, PtdIns(5)P-4-kinase isoform 2-alpha, type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform | |
| Full length human recombinant protein of human PIP4K2A (NP_005019) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction