Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIP5KL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563272
Description
Immunogen sequence: ISERYDIKGC EVSRWVDPAP EGSPLVLVLK DLNFQGKTIN LGPQRSWFLR QMELDTTFLR ELNVLDYSLL IAFQRLHE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
PIP5KL1 is a phosphoinositide kinase-like protein that lacks intrinsic lipid kinase activity but associates with type I PIPKs and may play a role in localization of PIPK activity (Chang et al., 2004 ).
Specifications
PIP5KL1 | |
Polyclonal | |
Unconjugated | |
PIP5KL1 | |
bA203J24.5; BC028795; Phosphatidylinositol 4-phosphate 5-kinase-like protein 1; phosphatidylinositol phosphate kinase homolog; phosphatidylinositol-4-phosphate 5-kinase like 1; phosphatidylinositol-4-phosphate 5-kinase-like 1; phosphatidylinositol-4-phosphate 5-kinase-like protein 1; PI(4)P 5-kinase-like protein 1; Pip5kl1; PIPKH; ptdIns(4)P-5-kinase-like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
138429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T9C9 | |
PIP5KL1 | |
Recombinant protein corresponding to Human PIP5KL1. Recombinant protein control fragment (Product #RP-93054). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction