Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5KL1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528615 Supplier Thermo Scientific Supplier No. PA528615

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3. Predicted reactivity: Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PIP5KL1 is a phosphoinositide kinase-like protein that lacks intrinsic lipid kinase activity but associates with type I PIPKs and may play a role in localization of PIPK activity (Chang et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5KL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PIP5KL1
Gene Accession No. Q5T9C9
Gene Alias bA203J24.5; BC028795; Phosphatidylinositol 4-phosphate 5-kinase-like protein 1; phosphatidylinositol phosphate kinase homolog; phosphatidylinositol-4-phosphate 5-kinase like 1; phosphatidylinositol-4-phosphate 5-kinase-like 1; phosphatidylinositol-4-phosphate 5-kinase-like protein 1; PI(4)P 5-kinase-like protein 1; Pip5kl1; PIPKH; ptdIns(4)P-5-kinase-like protein 1
Gene Symbols PIP5KL1
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 163 and 394 of Human PIP5KL1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138429
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.