Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5KL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980284
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0284 Supplier Bioss Supplier No. BS0451R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIP5KL1 is a phosphoinositide kinase-like protein that lacks intrinsic lipid kinase activity but associates with type I PIPKs and may play a role in localization of PIPK activity (Chang et al., 2004 ).

Specifications

Antigen PIP5KL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PIP5KL1
Gene Accession No. Q5T9C9
Gene Alias bA203J24.5; BC028795; Phosphatidylinositol 4-phosphate 5-kinase-like protein 1; Phosphatidylinositol phosphate kinase homolog; phosphatidylinositol-4-phosphate 5-kinase like 1; phosphatidylinositol-4-phosphate 5-kinase-like 1; phosphatidylinositol-4-phosphate 5-kinase-like protein 1; PI(4)P 5-kinase-like protein 1; PIP5KL1; Pipkh; ptdIns(4)P-5-kinase-like protein 1
Gene Symbols PIP5KL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PIP5KL1/PIPKH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138429
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.