Learn More
Description
PIPE 3297 dihydrochloride induces differentiation of oligodendrocyte progenitor cells (OPC) in vitro and in vivo, leading to significant increase of mature oligodendrocytes (OL) in healthy mice and inducing myelination. It also reduces disease score in mouse experimental autoimmune encephalomyelitis (EAE) model. Exhibits low β-arrestin2 recruitment activity, avoding the sedating effects typically associated to other KOR agonists employed in demyelinating diseases.
PIPE 3297 dihydrochloride is brain penetrant and leads to 90% occupancy in CNS in vivo (mouse).
PIPE 3297 exhibits cardiotoxity with 72% hERG inhibition (3 μM) and unstability in liver microsomes.
Specifications
Specifications
| Chemical Name or Material | 3-(2-((1-Benzylazetidin-3-yl)(cyclobutylmethyl)amino)ethyl)phenol dihydrochloride |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | KOR |
| Molecular Formula | C23H30N2O.2HCl |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
