Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pipecolic acid oxidase Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299598IR
Description
Pipecolic acid oxidase Polyclonal specifically detects Pipecolic acid oxidase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pipecolic acid oxidase | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 1.5.3.1, EC 1.5.3.7, L-pipecolate oxidase, L-pipecolic acid oxidase, LPIPOXperoxisomal sarcosine oxidase, pipecolic acid oxidase, PSO | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Pipecolic acid oxidase. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cell Biology, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
51268 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction