Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine, 99%, extra pure

Catalog No. p-185413
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg

Catalog No. AC131291000

Supplier: Thermo Scientific Chemicals 131291000

Only null left
Add to Cart

CAS: 110-85-0 | C4H10N2 | 86.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00005953
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
Synonym diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol
PubChem CID 4837
ChEBI CHEBI:28568
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Melting Point 107.0°C to 111.0°C
Color White to Yellow
Boiling Point 146.0°C to 148.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 23,4
Fieser 04,392
Merck Index 15,7576
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol and glycols, soluble in alcohol, practically insoluble in diethyl ether
Formula Weight 86.14
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Flakes
Chemical Name or Material Piperazine, anhydrous, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mis
EINECSNumber 203-808-3
RTECSNumber TK7800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.