Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine hexahydrate, 98%

Catalog No. AC131305000
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC131305000

Supplier: Thermo Scientific Chemicals 131305000

Only null left
Add to Cart

CAS: 142-63-2 | C4H10N2·6H2O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-63-2
Molecular Formula C4H10N2·6H2O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00149389
InChI Key AVRVZRUEXIEGMP-UHFFFAOYSA-N
Synonym piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
PubChem CID 120181
IUPAC Name piperazine;hexahydrate
SMILES C1CNCCN1.O.O.O.O.O.O

Specifications

Melting Point 41.0°C to 45.0°C
Color White
Boiling Point 145.0°C to 156.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 23,4
Merck Index 14,7464
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
Formula Weight 194.23
Percent Purity 98%
Physical Form Adhering Crystals
Chemical Name or Material Piperazine hexahydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
RTECSNumber TM0850000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.