Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piperazine hexahydrate, ≥99% (TLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
200 g
1 kg
Description
Featured Industries: Pharmaceutical (small molecule)

Specifications
Specifications
CAS | 142-63-2 |
Vapor Pressure | 0.8 mmHg (20°C) |
Flash Point | 88°C |
Physical Form | Neat |
Linear Formula | C4H10N2 · 6H2 O |
MDL Number | MFCD00149389 |
Melting Point | 42°C to 44°C |
Boiling Point | 145°C to 156°C |
Type | Analytical Standard |
Quantity | 200 g |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction